missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Tribromobenzene 95.0+%, TCI America™
$98.79 - $285.21
Chemical Identifiers
| CAS | 615-54-3 |
|---|---|
| Molecular Formula | C6H3Br3 |
| Molecular Weight (g/mol) | 314.802 |
| MDL Number | MFCD00000062 |
| InChI Key | FWAJPSIPOULHHH-UHFFFAOYSA-N |
| PubChem CID | 12002 |
| IUPAC Name | 1,2,4-tribromobenzene |
| SMILES | C1=CC(=C(C=C1Br)Br)Br |
Chemical Identifiers
| 615-54-3 | |
| 314.802 | |
| FWAJPSIPOULHHH-UHFFFAOYSA-N | |
| 1,2,4-tribromobenzene |
| C6H3Br3 | |
| MFCD00000062 | |
| 12002 | |
| C1=CC(=C(C=C1Br)Br)Br |
Specifications
| 615-54-3 | |
| White-Yellow | |
| MFCD00000062 | |
| benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2 | |
| C1=CC(=C(C=C1Br)Br)Br | |
| 314.802 | |
| 314.80 | |
| Crystalline Lumps |
| 42°C | |
| C6H3Br3 | |
| 5 g | |
| FWAJPSIPOULHHH-UHFFFAOYSA-N | |
| 1,2,4-tribromobenzene | |
| 12002 | |
| ≥95.0% (GC) | |
| 1,2,4-Tribromobenzene |
Safety and Handling
RTECSNumber : DC1956500
TSCA : Yes