missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2:5,6-Di-O-isopropylidene-D-mannitol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1212706

 View more versions of this product

Catalog No. AAL1212706


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2:5,6-Di-O-isopropylidene-D-mannitol
120°C to 122°C
MFCD00003211
83765
ODYBCPSCYHAGHA-ZYUZMQFOSA-N
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C
262.302
262.31
1707-77-3
C12H22O6
5 g
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
+6° (c=5 in Chloroform)
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
96011

Chemical Identifiers

1707-77-3
262.302
ODYBCPSCYHAGHA-ZYUZMQFOSA-N
96011
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C
C12H22O6
MFCD00003211
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
Safety and Handling

Safety and Handling

EINECSNumber : 216-954-8

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only