missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2,5-Pentanetriol 97.0+%, TCI America™
$268.76 - $673.56
Chemical Identifiers
CAS | 14697-46-2 |
---|---|
Molecular Formula | C5H12O3 |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD01709258 |
InChI Key | WEAYWASEBDOLRG-UHFFFAOYNA-N |
PubChem CID | 85763 |
IUPAC Name | pentane-1,2,5-triol |
SMILES | OCCCC(O)CO |
Chemical Identifiers
14697-46-2 | |
120.15 | |
WEAYWASEBDOLRG-UHFFFAOYNA-N | |
pentane-1,2,5-triol |
C5H12O3 | |
MFCD01709258 | |
85763 | |
OCCCC(O)CO |
Specifications
14697-46-2 | |
195°C | |
MFCD01709258 | |
WEAYWASEBDOLRG-UHFFFAOYNA-N | |
pentane-1,2,5-triol | |
85763 | |
≥97.0% (GC) | |
1,2,5-Pentanetriol |
Yellow | |
C5H12O3 | |
1 g | |
OCCCC(O)CO | |
120.15 | |
120.15 | |
Liquid |
Safety and Handling
RTECSNumber : SA3327000
TSCA : No