missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,6-Hexanetriol, 97+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 139831000

 View more versions of this product

Catalog No. AC139831000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 2, 6-Hexanetriol
106-69-4
100.0
Yellow
4.0 to 7.0 (30% soln. at 20°C)
198°C
97% min. (GC)
C6H14O3
HO(CH2)4CH(OH)CH2OH
100 g
1.109
Solubility in water: miscible with water. Other solubilities: miscible with acetone and alcohol, immiscible with benzene, toluene, heptane and, diethyl ether, practically insoluble in ether, low solubility in non-polar solvents
OCCCCC(O)CO
134.18
134.18
Extra Pure
97%
97.0
25.0°C to 32.0°C
1.1090g/mL
178.0°C (6.7 mmHg)
Authentic
Glass bottle
1.4750 to 1.4770
MFCD00002976
01,IV,2784
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
ZWVMLYRJXORSEP-UHFFFAOYNA-N
hexane-1,2,6-triol
7823
97+%
Viscous Liquid

Chemical Identifiers

106-69-4
134.18
ZWVMLYRJXORSEP-UHFFFAOYNA-N
7823
OCCCCC(O)CO
C6H14O3
MFCD00002976
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
hexane-1,2,6-triol
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 203-424-6

RTECSNumber : MO4650000

TSCA : TSCA

Recommended Storage : the product may solidify at ca. 32°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only