Learn More
1,2-Benzanthracene, 99%
CAS: 56-55-3 | C18H12 | 228.29 g/mol
$251.66 - $251.66
Chemical Identifiers
| CAS | 56-55-3 |
|---|---|
| Molecular Formula | C18H12 |
| Molecular Weight (g/mol) | 228.29 |
| MDL Number | MFCD00003599 |
| InChI Key | DXBHBZVCASKNBY-UHFFFAOYSA-N |
| Synonym | benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene |
| PubChem CID | 5954 |
| ChEBI | CHEBI:51348 |
| IUPAC Name | benzo[a]anthracene |
| SMILES | C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC105250010
|
Thermo Scientific Chemicals
105250010 |
1 g | Glass bottle |
Each for $251.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56-55-3 | |
| 228.29 | |
| DXBHBZVCASKNBY-UHFFFAOYSA-N | |
| 5954 | |
| benzo[a]anthracene |
| C18H12 | |
| MFCD00003599 | |
| benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene | |
| CHEBI:51348 | |
| C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 |
Specifications
| 56-55-3 | |
| 100.0 | |
| Beige to Green-Yellow | |
| Authentic | |
| Glass bottle | |
| MFCD00003599 | |
| 05,718 | |
| benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene | |
| DXBHBZVCASKNBY-UHFFFAOYSA-N | |
| benzo[a]anthracene | |
| 5954 | |
| 228.29 | |
| Crystalline Powder |
| 98.5 | |
| 158°C to 161°C | |
| 437.6°C | |
| 98.5% min. (GC) | |
| C18H12 | |
| 1 g | |
| 15,1064 | |
| Solubility in water: insoluble in water. Other solubilities: difficulty soluble in boiling alcohol,soluble in most other common organic solvents | |
| C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1 | |
| 228.29 | |
| CHEBI:51348 | |
| 99% | |
| 1, 2-Benzanthracene, 99% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-280-6
RTECSNumber : CV9275000
TSCA : TSCA
RUO – Research Use Only