missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzisothiazol-3(2H)-one 98.0+%, TCI America™
$152.86 - $1330.94
Chemical Identifiers
| CAS | 2634-33-5 |
|---|---|
| Molecular Formula | C7H5NOS |
| Molecular Weight (g/mol) | 151.183 |
| MDL Number | MFCD00127753 |
| InChI Key | DMSMPAJRVJJAGA-UHFFFAOYSA-N |
| Synonym | 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one |
| PubChem CID | 17520 |
| ChEBI | CHEBI:167099 |
| IUPAC Name | 1,2-benzothiazol-3-one |
| SMILES | C1=CC=C2C(=C1)C(=O)NS2 |
Chemical Identifiers
| 2634-33-5 | |
| 151.183 | |
| DMSMPAJRVJJAGA-UHFFFAOYSA-N | |
| 17520 | |
| 1,2-benzothiazol-3-one |
| C7H5NOS | |
| MFCD00127753 | |
| 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one | |
| CHEBI:167099 | |
| C1=CC=C2C(=C1)C(=O)NS2 |
Specifications
| 2634-33-5 | |
| C7H5NOS | |
| 25 g | |
| 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one | |
| C1=CC=C2C(=C1)C(=O)NS2 | |
| 151.183 | |
| CHEBI:167099 | |
| ≥98.0% (GC) | |
| 1,2-Benzisothiazol-3(2H)-one |
| 158°C | |
| MFCD00127753 | |
| 3077 | |
| DMSMPAJRVJJAGA-UHFFFAOYSA-N | |
| 1,2-benzothiazol-3-one | |
| 17520 | |
| 151.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1845
RTECSNumber : DE4620000
TSCA : Yes