Learn More
1,2-Benzisoxazole, 95%, Thermo Scientific Chemicals
$70.48 - $224.62
Chemical Identifiers
CAS | 271-95-4 |
---|---|
Molecular Formula | C7H5NO |
Molecular Weight (g/mol) | 119.123 |
MDL Number | MFCD00005852 |
InChI Key | KTZQTRPPVKQPFO-UHFFFAOYSA-N |
Synonym | 1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine |
PubChem CID | 71073 |
ChEBI | CHEBI:51554 |
IUPAC Name | 1,2-benzoxazole |
SMILES | C1=CC=C2C(=C1)C=NO2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2340003
|
Thermo Scientific Chemicals
B2340003 |
1 g |
Each for $70.48
|
|
|||||
AAB2340006
|
Thermo Scientific Chemicals
B2340006 |
5 g |
Each for $224.62
|
|
|||||
Description
1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water. 4.1 mg/mL in water at 25°C
Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
Chemical Identifiers
271-95-4 | |
119.123 | |
KTZQTRPPVKQPFO-UHFFFAOYSA-N | |
71073 | |
1,2-benzoxazole |
C7H5NO | |
MFCD00005852 | |
1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine | |
CHEBI:51554 | |
C1=CC=C2C(=C1)C=NO2 |
Specifications
271-95-4 | |
90°C to 92°C (15 mmHg) | |
C7H5NO | |
MFCD00005852 | |
2154 | |
1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine | |
KTZQTRPPVKQPFO-UHFFFAOYSA-N | |
1,2-benzoxazole | |
71073 | |
119.12 | |
1,2-Benzisoxazole |
1.169 | |
86°C (186°F) | |
1.562 | |
1 g | |
Light sensitive | |
Soluble in water. 4.1mg/mL in water at 25°C | |
C1=CC=C2C(=C1)C=NO2 | |
119.123 | |
CHEBI:51554 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 205-983-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only