Learn More
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals
$519.88 - $519.88
Chemical Identifiers
CAS | 228121-39-9 |
---|---|
Molecular Formula | C30H48BF4P2Rh |
Molecular Weight (g/mol) | 660.37 |
MDL Number | MFCD01862464 |
InChI Key | DSYBEQKPSKLNMC-RTXTXWNLNA-N |
PubChem CID | 11549211 |
IUPAC Name | cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate |
SMILES | [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC435182500
|
Thermo Scientific Chemicals
435182500 |
250 mg |
Each for $519.88
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
228121-39-9 | |
660.37 | |
DSYBEQKPSKLNMC-RTXTXWNLNA-N | |
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate |
C30H48BF4P2Rh | |
MFCD01862464 | |
11549211 | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC |
Specifications
228121-39-9 | |
100.0 | |
52.4 to 56.8% C | |
97% | |
MFCD01862464 | |
DSYBEQKPSKLNMC-RTXTXWNLNA-N | |
cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate | |
11549211 | |
97% | |
1,2-Bis((2R,5R)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate |
96.0 | |
Brown to Orange | |
Authentic | |
C30H48BF4P2Rh | |
250 mg | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC | |
660.37 | |
660.37 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning
RUO – Research Use Only