missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 435202500

 View more versions of this product

Catalog No. AC435202500


Explore available promotions

Description

Description

Specifications

Specifications

1,2-Bis((2R,5R)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
96.0
Brown to Yellow
Authentic
C34H56BF4P2Rh
250 mg
AXLKBYDAACXFBH-KFORCWLUNA-N
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide
12964416
97%
569650-64-2
100.0
54.7 to 59.3% C
97%
MFCD07369039
1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C
716.48
716.48
Powder

Chemical Identifiers

569650-64-2
716.48
AXLKBYDAACXFBH-KFORCWLUNA-N
12964416
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C
C34H56BF4P2Rh
MFCD07369039
1,2-bis 2r,5r-2,5-diisopropylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate, r,r-i-pr-duphos-rh, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene 1,5-cyclooctadiene rhodium i tetrafluoroborate
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only