missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

$261.52 - $571.57

Chemical Identifiers

CAS 213343-64-7
Molecular Formula C30H48BF4P2Rh
Molecular Weight (g/mol) 660.37
MDL Number MFCD01862465
InChI Key DSYBEQKPSKLNMC-RTXTXWNLNA-N
Synonym s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate
PubChem CID 11006841
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC435191000
SDS View Documents
Thermo Scientific Chemicals
435191000
100 mg Glass bottle
Each for $261.52
 
AC435192500
SDS View Documents
Thermo Scientific Chemicals
435192500
250 mg Glass bottle
Each for $571.57
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

213343-64-7
660.37
DSYBEQKPSKLNMC-RTXTXWNLNA-N
11006841
C30H48BF4P2Rh
MFCD01862465
s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC

Specifications

213343-64-7
100.0
52.4 to 56.8% C
97%
C30H48BF4P2Rh
100 mg
DSYBEQKPSKLNMC-RTXTXWNLNA-N
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl}-2,5-diethylphospholane tetrafluoroboranuide
11006841
97%
1,2-Bis((2S,5S)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
96.0
Brown to Orange
Authentic
Glass bottle
MFCD01862465
s,s-et-duphos-rh, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium i tetrafluoroborate, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene cyclooctadiene rhodium l tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC
660.37
660.37
Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only