missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals
$299.49 - $933.18
Chemical Identifiers
CAS | 528854-34-4 |
---|---|
Molecular Formula | C22H44P2 |
Molecular Weight (g/mol) | 370.542 |
MDL Number | MFCD08705244 |
InChI Key | IRVIFEWWPYKALC-GXRSIYKFSA-N |
Synonym | 1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan |
PubChem CID | 11707202 |
IUPAC Name | (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane |
SMILES | CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH60017MD
|
Thermo Scientific Chemicals
H60017MD |
250 mg |
Each for $299.49
|
|
||||||
AAH6001703
|
Thermo Scientific Chemicals
H6001703 |
1 g |
Each for $933.18
|
|
||||||
Description
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air sensitive. Keep away from strong oxidizing agents.
Chemical Identifiers
528854-34-4 | |
370.542 | |
IRVIFEWWPYKALC-GXRSIYKFSA-N | |
11707202 | |
CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C |
C22H44P2 | |
MFCD08705244 | |
1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan | |
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane |
Specifications
528854-34-4 | |
C22H44P2 | |
250 mg | |
1,2-bis 2s,5s-2,5-di-i-propylphospholano ethan | |
IRVIFEWWPYKALC-GXRSIYKFSA-N | |
(2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]ethyl]-2,5-di(propan-2-yl)phospholane | |
11707202 | |
≥97% |
67°C to 72°C | |
MFCD08705244 | |
Air Sensitive | |
Insoluble in water. | |
CC(C)C1CCC(P1CCP2C(CCC2C(C)C)C(C)C)C(C)C | |
370.542 | |
370.53 | |
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only