Learn More
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals
$1019.50 - $1019.50
Chemical Identifiers
CAS | 213343-65-8 |
---|---|
Molecular Formula | C22H40BF4P2Rh |
Molecular Weight (g/mol) | 556.22 |
MDL Number | MFCD09038435 |
InChI Key | CVUPEPGZCZHSOF-BDNLEJQTNA-N |
Synonym | 1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate |
PubChem CID | 12964417 |
SMILES | [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC435235000
|
Thermo Scientific Chemicals
435235000 |
500 mg |
Each for $1,019.50
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
213343-65-8 | |
556.22 | |
CVUPEPGZCZHSOF-BDNLEJQTNA-N | |
12964417 |
C22H40BF4P2Rh | |
MFCD09038435 | |
1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C |
Specifications
213343-65-8 | |
100.0 | |
Brown to Orange | |
97% | |
MFCD09038435 | |
1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene rhodium i tetrafluoroborate, s,s-me-bpe-rh, --1,2-bis 2s,5s-2,5-dimethylphospholano ethane cyclooctadiene-rhodium i tetrafluoroborate | |
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C | |
556.22 | |
556.21 | |
Crystalline Powder |
96.0 | |
152.0°C to 156.0°C | |
Authentic | |
C22H40BF4P2Rh | |
500 mg | |
CVUPEPGZCZHSOF-BDNLEJQTNA-N | |
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl}-2,5-dimethylphospholane tetrafluoroboranuide | |
12964417 | |
97% | |
1,2-Bis((2S,5S)-2,5-Dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only