missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 363230050

 View more versions of this product

Catalog No. AC363230050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride
98.5
263.0°C to 265.0°C
98.5% min.
99%
C26H24Cl2NiP2
5 g
Solubility in water: insoluble in water
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
528.02
528.02
Crystalline Powder
14647-23-5
100.0
Orange to Red
Authentic
Glass bottle
MFCD00013313
1,2-bis diphenylphosphino ethane nickel ii chloride
XXECWTBMGGXMKP-UHFFFAOYSA-L
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
131664338
99%

Chemical Identifiers

14647-23-5
528.02
XXECWTBMGGXMKP-UHFFFAOYSA-L
131664338
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C26H24Cl2NiP2
MFCD00013313
1,2-bis diphenylphosphino ethane nickel ii chloride
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only