missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride 96.0+%, TCI America™
$46.44 - $396.51
Chemical Identifiers
CAS | 14647-23-5 |
---|---|
Molecular Formula | C26H24Cl2NiP2 |
Molecular Weight (g/mol) | 528.02 |
MDL Number | MFCD00013313 |
InChI Key | XXECWTBMGGXMKP-UHFFFAOYSA-L |
Synonym | 1,2-bis diphenylphosphino ethane nickel ii chloride |
PubChem CID | 131664338 |
IUPAC Name | [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel |
SMILES | Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
14647-23-5 | |
528.02 | |
XXECWTBMGGXMKP-UHFFFAOYSA-L | |
131664338 | |
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C26H24Cl2NiP2 | |
MFCD00013313 | |
1,2-bis diphenylphosphino ethane nickel ii chloride | |
[2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel |
Specifications
14647-23-5 | |
MFCD00013313 | |
1,2-bis diphenylphosphino ethane nickel ii chloride | |
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
528.02 | |
528.02 | |
Crystalline Powder |
C26H24Cl2NiP2 | |
1 g | |
XXECWTBMGGXMKP-UHFFFAOYSA-L | |
[2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel | |
131664338 | |
≥96.0% (T) | |
[1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride |
Safety and Handling
TSCA : No