missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis(methanesulfonamido)benzene, 97%, Thermo Scientific Chemicals
$55.15 - $142.62
Chemical Identifiers
CAS | 7596-80-7 |
---|---|
Molecular Formula | C8H12N2O4S2 |
Molecular Weight (g/mol) | 264.31 |
MDL Number | MFCD00276605 |
InChI Key | BYZJRLRCNOECEV-UHFFFAOYSA-N |
Synonym | 1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide |
PubChem CID | 227307 |
IUPAC Name | N-[2-(methanesulfonamido)phenyl]methanesulfonamide |
SMILES | CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1983088
|
Thermo Scientific Chemicals
A1983088 |
2.5 g |
Each for $55.15
|
|
|||||
AAA1983009
|
Thermo Scientific Chemicals
A1983009 |
10 g |
Each for $142.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7596-80-7 | |
264.31 | |
BYZJRLRCNOECEV-UHFFFAOYSA-N | |
227307 | |
CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O |
C8H12N2O4S2 | |
MFCD00276605 | |
1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide | |
N-[2-(methanesulfonamido)phenyl]methanesulfonamide |
Specifications
7596-80-7 | |
Odorless | |
MFCD00276605 | |
1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide | |
CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O | |
264.31 | |
264.32 | |
1,2-Bis(methanesulfonamido)benzene |
211°C to 214°C | |
C8H12N2O4S2 | |
2.5 g | |
BYZJRLRCNOECEV-UHFFFAOYSA-N | |
N-[2-(methanesulfonamido)phenyl]methanesulfonamide | |
227307 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only