missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Butanediol 98.0+%, TCI America™
$35.41 - $123.03
Chemical Identifiers
| CAS | 584-03-2 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.12 |
| MDL Number | MFCD00004570 |
| InChI Key | BMRWNKZVCUKKSR-UHFFFAOYNA-N |
| Synonym | 1,2-butanediol, 1,2-butylene glycol, 1,2-dihydroxybutane, alpha-butyleneglycol, alpha-butylene glycol, 1,2-butandiol, 1,2-dihydroxy butane, butanediol, 1,2, .alpha.-butylene glycol, 1,2-butylene-glycol |
| PubChem CID | 11429 |
| ChEBI | CHEBI:52682 |
| IUPAC Name | butane-1,2-diol |
| SMILES | CCC(O)CO |
Chemical Identifiers
| 584-03-2 | |
| 90.12 | |
| BMRWNKZVCUKKSR-UHFFFAOYNA-N | |
| 11429 | |
| butane-1,2-diol |
| C4H10O2 | |
| MFCD00004570 | |
| 1,2-butanediol, 1,2-butylene glycol, 1,2-dihydroxybutane, alpha-butyleneglycol, alpha-butylene glycol, 1,2-butandiol, 1,2-dihydroxy butane, butanediol, 1,2, .alpha.-butylene glycol, 1,2-butylene-glycol | |
| CHEBI:52682 | |
| CCC(O)CO |
Specifications
| 584-03-2 | |
| 194°C | |
| MFCD00004570 | |
| 1,2-butanediol, 1,2-butylene glycol, 1,2-dihydroxybutane, alpha-butyleneglycol, alpha-butylene glycol, 1,2-butandiol, 1,2-dihydroxy butane, butanediol, 1,2, .alpha.-butylene glycol, 1,2-butylene-glycol | |
| CCC(O)CO | |
| 90.12 | |
| CHEBI:52682 | |
| ≥98.0% (GC) | |
| 1,2-Butanediol |
| Yellow | |
| C4H10O2 | |
| 25 g | |
| BMRWNKZVCUKKSR-UHFFFAOYNA-N | |
| butane-1,2-diol | |
| 11429 | |
| 90.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0235
RTECSNumber : EK0380000
TSCA : Yes