missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Chloro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™

$43.96 - $513.79

Chemical Identifiers

CAS 114878-60-3
Molecular Formula C10H12ClN3O2
Molecular Weight (g/mol) 241.675
MDL Number MFCD00640776
InChI Key JNUWBYPJOBSXDN-UHFFFAOYSA-N
PubChem CID 2771331
IUPAC Name 1-(2-chloro-4-nitrophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63232MD
View Documents
Thermo Scientific Chemicals
H63232MD
250 mg
Each for $43.96
 
AAH6323203
View Documents
Thermo Scientific Chemicals
H6323203
1 g
Each for $120.58
 
AAH6323206
View Documents
Thermo Scientific Chemicals
H6323206
5 g
Each for $513.79
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

114878-60-3
241.675
JNUWBYPJOBSXDN-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)piperazine
C10H12ClN3O2
MFCD00640776
2771331
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

114878-60-3
MFCD00640776
1-2-chloro-4-nitrophenyl piperazine, 2-chloro-4-nitrophenyl piperazine, 1-2-chloro-4-nitrophenyl-piperazine, piperazine, 1-2-chloro-4-nitrophenyl, 1-2-chloro-4-nitro-phenyl-piperazine, piperazine,1-2-chloro-4-nitrophenyl, pubchem9343, acmc-1bvwp, cambridge id 5668126, 1-2-chloro-4-nitro-phenyl piperazine
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
241.675
241.67
Solid
C10H12ClN3O2
250 mg
JNUWBYPJOBSXDN-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)piperazine
2771331
97%
1-(2-Chloro-4-nitrophenyl)piperazine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'