missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole 98.0+%, TCI America™

$150.98 - $443.65

Chemical Identifiers

CAS 628333-86-8
Molecular Formula C29H33N2P
Molecular Weight (g/mol) 440.571
MDL Number MFCD09038823
InChI Key JAIIBHBCNPAPPF-UHFFFAOYSA-N
Synonym TrippyPhos
PubChem CID 11430600
IUPAC Name ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane
SMILES CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D38341G
View Documents
TCI America
D38341G
1 g
Each for $150.98
Only null left
 
D38345G
View Documents
TCI America
D38345G
5 g
Each for $443.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

628333-86-8
440.571
JAIIBHBCNPAPPF-UHFFFAOYSA-N
11430600
CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C
C29H33N2P
MFCD09038823
TrippyPhos
ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane

Specifications

628333-86-8
White-Yellow
MFCD09038823
TrippyPhos
CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C
440.571
440.57
Crystalline Powder
142°C
C29H33N2P
1 g
JAIIBHBCNPAPPF-UHFFFAOYSA-N
ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane
11430600
≥98.0% (GC,T)
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'