missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Diamino-2-methylpropane 97.0+%, TCI America™
$207.62 - $852.94
Chemical Identifiers
CAS | 811-93-8 |
---|---|
Molecular Formula | C4H12N2 |
Molecular Weight (g/mol) | 88.154 |
MDL Number | MFCD00008054 |
InChI Key | OPCJOXGBLDJWRM-UHFFFAOYSA-N |
Synonym | 2-Methyl-1,2-propanediamine |
PubChem CID | 13128 |
IUPAC Name | 2-methylpropane-1,2-diamine |
SMILES | CC(C)(CN)N |
Chemical Identifiers
811-93-8 | |
88.154 | |
OPCJOXGBLDJWRM-UHFFFAOYSA-N | |
13128 | |
CC(C)(CN)N |
C4H12N2 | |
MFCD00008054 | |
2-Methyl-1,2-propanediamine | |
2-methylpropane-1,2-diamine |
Specifications
811-93-8 | |
52°C | |
MFCD00008054 | |
2734 | |
OPCJOXGBLDJWRM-UHFFFAOYSA-N | |
2-methylpropane-1,2-diamine | |
13128 | |
≥97.0% (GC,T) | |
1,2-Diamino-2-methylpropane |
Yellow | |
C4H12N2 | |
5 mL | |
2-Methyl-1,2-propanediamine | |
CC(C)(CN)N | |
88.154 | |
88.15 | |
Liquid |
Safety and Handling
RTECSNumber : TX8250000
TSCA : No