missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dibromo-3,3-dimethyl-1-butene 96.0+%, TCI America™

Supplier:  TCI America D291425G

Catalog No. D291425G


Explore available promotions

Description

Description

Specifications

Specifications

1,2-Dibromo-3,3-dimethyl-1-butene
Yellow
MFCD06797090
JDGBPINHTJNHMH-UHFFFAOYSA-N
1,2-dibromo-3,3-dimethylbut-1-ene
44630273
≥96.0% (GC)
99584-95-9
C6H10Br2
25 g
CC(C)(C)C(Br)=CBr
241.95
241.95
Liquid

Chemical Identifiers

99584-95-9
241.95
JDGBPINHTJNHMH-UHFFFAOYSA-N
1,2-dibromo-3,3-dimethylbut-1-ene
C6H10Br2
MFCD06797090
44630273
CC(C)(C)C(Br)=CBr
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions