missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3355806

 View more versions of this product

Catalog No. AAH3355806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-Dibromo-3,4,5,6-tetramethylbenzene
C10H12Br2
5 g
VEZJOZSIGSNYEB-UHFFFAOYSA-N
1,2-dibromo-3,4,5,6-tetramethylbenzene
2752606
98%
36321-73-0
MFCD00154987
benzene,1,2-dibromo-3,4,5,6-tetramethyl, pubchem21478, benzene, 1,2-dibromo-3,4,5,6-tetramethyl, 1,2-bis bromanyl-3,4,5,6-tetramethyl-benzene
CC1=C(C(=C(C(=C1C)Br)Br)C)C
292.014
292.01

Chemical Identifiers

36321-73-0
292.014
VEZJOZSIGSNYEB-UHFFFAOYSA-N
2752606
CC1=C(C(=C(C(=C1C)Br)Br)C)C
C10H12Br2
MFCD00154987
benzene,1,2-dibromo-3,4,5,6-tetramethyl, pubchem21478, benzene, 1,2-dibromo-3,4,5,6-tetramethyl, 1,2-bis bromanyl-3,4,5,6-tetramethyl-benzene
1,2-dibromo-3,4,5,6-tetramethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only