Learn More
1,2-Dibromobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1476514
Description
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.
Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
1,2-Dibromobenzene | |
4°C to 6°C | |
224°C to 226°C | |
Strong | |
1.611 | |
25 g | |
970241 | |
Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water. | |
C1=CC=C(C(=C1)Br)Br | |
235.906 | |
CHEBI:37152 | |
98% |
583-53-9 | |
1.985 | |
91°C (195°F) | |
C6H4Br2 | |
MFCD00000057 | |
UN3082 | |
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene | |
WQONPSCCEXUXTQ-UHFFFAOYSA-N | |
1,2-dibromobenzene | |
11414 | |
235.91 |
Chemical Identifiers
583-53-9 | |
235.906 | |
WQONPSCCEXUXTQ-UHFFFAOYSA-N | |
11414 | |
1,2-dibromobenzene |
C6H4Br2 | |
MFCD00000057 | |
o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene | |
CHEBI:37152 | |
C1=CC=C(C(=C1)Br)Br |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 209-507-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only