missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dibromobutane 98.0+%, TCI America™
Supplier: TCI America D017425G
Specifications
| 1,2-Dibromobutane | |
| Yellow | |
| C4H8Br2 | |
| 25 g | |
| CZWSZZHGSNZRMW-UHFFFAOYNA-N | |
| 1,2-dibromobutane | |
| 10792 | |
| ≥98.0% (GC) |
| 533-98-2 | |
| 166°C | |
| MFCD00000157 | |
| butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide | |
| CCC(Br)CBr | |
| 215.92 | |
| 215.92 | |
| Liquid |
Chemical Identifiers
| 533-98-2 | |
| 215.92 | |
| CZWSZZHGSNZRMW-UHFFFAOYNA-N | |
| 10792 | |
| CCC(Br)CBr |
| C4H8Br2 | |
| MFCD00000157 | |
| butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide | |
| 1,2-dibromobutane |
Safety and Handling
TSCA : Yes