missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dibromoethane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 112790010

 View more versions of this product

Catalog No. AC112790010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-Dibromoethane
98.5
9.0°C to 10.0°C
2.1730g/mL
>104°C
98.5% min. (GC)
C2H4Br2
BrCH2CH2Br
01,9
2.173
ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
PAAZPARNPHGIKF-UHFFFAOYSA-N
1,2-dibromoethane
7839
187.86
Liquid
106-93-4
100.0
Colorless to Yellow
131.0°C to 132.0°C
Authentic
Glass bottle
1.5375 to 1.5395
1 L
01,374; 04,126; 05,187; 12,570
15,385
Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents
C(CBr)Br
187.86
CHEBI:28534
99%

Chemical Identifiers

106-93-4
187.86
ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom
CHEBI:28534
C(CBr)Br
C2H4Br2
PAAZPARNPHGIKF-UHFFFAOYSA-N
7839
1,2-dibromoethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause cancer.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-444-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only