Learn More
1,2-Dichlorobutane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2130706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2-Dichlorobutane | |
1.112 | |
30°C (86°F) | |
1.445 | |
5 g | |
1731376 | |
PQBOTZNYFQWRHU-UHFFFAOYNA-N | |
1,2-dichlorobutane | |
12017 | |
≥97% |
616-21-7 | |
124°C | |
C4H8Cl2 | |
MFCD00013676 | |
UN1993 | |
butane, 1,2-dichloro, butane,2-dichloro, 1,2-butylene chloride, acmc-1b1ry, dsstox_cid_7244, dsstox_rid_78366, dsstox_gsid_27244, 1,2-dichlorobutane, pqbotznyfqwrhu-uhfffaoysa | |
CCC(Cl)CCl | |
127.01 | |
127.01 |
Chemical Identifiers
616-21-7 | |
127.01 | |
PQBOTZNYFQWRHU-UHFFFAOYNA-N | |
12017 | |
CCC(Cl)CCl |
C4H8Cl2 | |
MFCD00013676 | |
butane, 1,2-dichloro, butane,2-dichloro, 1,2-butylene chloride, acmc-1b1ry, dsstox_cid_7244, dsstox_rid_78366, dsstox_gsid_27244, 1,2-dichlorobutane, pqbotznyfqwrhu-uhfffaoysa | |
1,2-dichlorobutane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 210-469-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only