missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 278740050

 View more versions of this product

Catalog No. AC278740050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 2-Dimethyl-3-hydroxy-4-pyridone
99.0
271°C to 273°C
6 to 8 (1% soln.)
Authentic
Glass bottle
MFCD00134497
15, 2862
Solubility in water: soluble in hot water. Other solubilities: soluble in dmso
CC1=C(C(=O)C=CN1C)O
139.15
CHEBI:68554
99+%
30652-11-0
100.0
White
0.5% max.
99+%
C7H9NO2
5 g
deferiprone, ferriprox, 3-hydroxy-1,2-dimethyl-4 1h-pyridone, 3-hydroxy-1,2-dimethylpyridin-4 1h-one, 1,2-dimethyl-3-hydroxy-4-pyridone, 1,2-dimethyl-3-hydroxypyrid-4-one, 1,2-dimethyl-3-hydroxypyridine-4-one, cp20 chelating agent, unii-2bty8kh53l, ferriprox tn
TZXKOCQBRNJULO-UHFFFAOYSA-N
3-hydroxy-1,2-dimethylpyridin-4-one
2972
139.15
Crystalline Powder or Needles

Chemical Identifiers

30652-11-0
139.15
TZXKOCQBRNJULO-UHFFFAOYSA-N
2972
3-hydroxy-1,2-dimethylpyridin-4-one
C7H9NO2
MFCD00134497
deferiprone, ferriprox, 3-hydroxy-1,2-dimethyl-4 1h-pyridone, 3-hydroxy-1,2-dimethylpyridin-4 1h-one, 1,2-dimethyl-3-hydroxy-4-pyridone, 1,2-dimethyl-3-hydroxypyrid-4-one, 1,2-dimethyl-3-hydroxypyridine-4-one, cp20 chelating agent, unii-2bty8kh53l, ferriprox tn
CHEBI:68554
CC1=C(C(=O)C=CN1C)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only