missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dimethyl-5-nitroimidazole, 98%, Thermo Scientific™

Chemical Identifiers

CAS 551-92-8
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00047046
InChI Key IBXPYPUJPLLOIN-UHFFFAOYSA-N
Synonym dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
PubChem CID 3090
IUPAC Name 1,2-dimethyl-5-nitroimidazole
SMILES CN1C(C)=NC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210331000
SDS View Documents
Thermo Scientific Chemicals
210331000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210330010
SDS View Documents
Thermo Scientific Chemicals
210330010
1kg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

551-92-8
141.13
IBXPYPUJPLLOIN-UHFFFAOYSA-N
3090
CN1C(C)=NC=C1[N+]([O-])=O
C5H7N3O2
MFCD00047046
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
1,2-dimethyl-5-nitroimidazole

Specifications

551-92-8
100.0
97.5% min. (HPLC)
C5H7N3O2
100g
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
CN1C(C)=NC=C1[N+]([O-])=O
141.13
141.13
1, 2-Dimethyl-5-nitroimidazole
97.5
Authentic
Glass bottle
MFCD00047046
15,329
IBXPYPUJPLLOIN-UHFFFAOYSA-N
1,2-dimethyl-5-nitroimidazole
3090
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-001-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only