Learn More
1,2-Epoxy-3-methylbutane, 98+%, Thermo Scientific Chemicals
$123.10 - $522.29
Chemical Identifiers
CAS | 1438-14-8 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.134 |
MDL Number | MFCD02683501 |
InChI Key | REYZXWIIUPKFTI-UHFFFAOYSA-N |
Synonym | 2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns |
PubChem CID | 102618 |
IUPAC Name | 2-propan-2-yloxirane |
SMILES | CC(C)C1CO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1929103
|
Thermo Scientific Chemicals
L1929103 |
1 g |
Each for $123.10
|
|
|||||
AAL1929106
|
Thermo Scientific Chemicals
L1929106 |
5 g |
Each for $522.29
|
|
|||||
Description
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
Chemical Identifiers
1438-14-8 | |
86.134 | |
REYZXWIIUPKFTI-UHFFFAOYSA-N | |
102618 | |
CC(C)C1CO1 |
C5H10O | |
MFCD02683501 | |
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns | |
2-propan-2-yloxirane |
Specifications
1438-14-8 | |
72°C to 75°C | |
Recognizable | |
1.3915 | |
1 g | |
102418 | |
2-isopropyloxirane, 1,2-epoxy-3-methylbutane, isopropyloxirane, oxirane, 1-methylethyl, butane, 1,2-epoxy-3-methyl, 2-propan-2-yl oxirane, oxirane,2-1-methylethyl, 1-methylethyl-oxirane, isopropylethylene oxide, acmc-1asns | |
REYZXWIIUPKFTI-UHFFFAOYSA-N | |
2-propan-2-yloxirane | |
102618 | |
≥98% |
0.815 | |
−12°C (10°F) | |
C5H10O | |
MFCD02683501 | |
UN1993 | |
Moisture sensitive | |
Not miscible or difficult to mix in water. | |
CC(C)C1CO1 | |
86.134 | |
86.13 | |
1,2-Epoxy-3-methylbutane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 215-869-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only