missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 98.0+%, TCI America™
Supplier: TCI America E0956250ML
Specifications
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) | |
-100°C | |
159°C | |
MFCD00022356 | |
3271 | |
SLJFKNONPLNAPF-UHFFFAOYSA-N | |
4-ethenyl-7-oxabicyclo[4.1.0]heptane | |
7832 | |
≥98.0% (GC) |
106-86-5 | |
Colorless | |
C8H12O | |
250 mL | |
4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane | |
C=CC1CCC2C(C1)O2 | |
124.183 | |
124.18 | |
Liquid |
Chemical Identifiers
106-86-5 | |
124.183 | |
SLJFKNONPLNAPF-UHFFFAOYSA-N | |
7832 | |
C=CC1CCC2C(C1)O2 |
C8H12O | |
MFCD00022356 | |
4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane | |
4-ethenyl-7-oxabicyclo[4.1.0]heptane |
Safety and Handling
EINECSNumber : (3)-2296
RTECSNumber : RN8770000
TSCA : Yes
Recommended Storage : Refrigerator