missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals SB01820EA

 View more versions of this product

Catalog No. SB01820EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
97.0
Amber glass bottle
FC6H4C6H4C(O)CH2CH3
10g
ZXIQZWXVSBJPTC-UHFFFAOYSA-N
1-[4-(2-fluorophenyl)phenyl]propan-1-one
2737490
97%
37989-92-7
97%
C15H13FO
MFCD00236612
1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenyl
CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F
228.266
228.26

Chemical Identifiers

37989-92-7
228.266
ZXIQZWXVSBJPTC-UHFFFAOYSA-N
2737490
CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F
C15H13FO
MFCD00236612
1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenyl
1-[4-(2-fluorophenyl)phenyl]propan-1-one
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions