missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(2-Fluorophenyl)ethanol, 95%, Thermo Scientific Chemicals
$185.03 - $185.03
Chemical Identifiers
CAS | 445-26-1 |
---|---|
Molecular Formula | C8H9FO |
Molecular Weight (g/mol) | 140.157 |
MDL Number | MFCD00014401 |
InChI Key | SXFYVXSOEBCFLV-UHFFFAOYSA-N |
Synonym | 1-2-fluorophenyl ethanol, 2-fluorophenylmethylcarbinol, 2-fluoro-alpha-methylbenzyl alcohol, 1-2-fluorophenyl ethan-1-ol, o-fluorophenylmethylcarbinol, 2-fluorophenyl methyl carbinol, 2-fluorophenylethanol, 1-m-fluorophenyl ethanol, 1-2'-fluorophenyl ethanol, fluorophenylmethylcarbinol |
PubChem CID | 123066 |
IUPAC Name | 1-(2-fluorophenyl)ethanol |
SMILES | CC(C1=CC=CC=C1F)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3190009
|
Thermo Scientific Chemicals
H3190009 |
10 g |
Each for $185.03
|
|
|||||
Description
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material in organic synthesis, agricultural fields and in dyestuffs.
Solubility
Immiscible in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
Chemical Identifiers
445-26-1 | |
140.157 | |
SXFYVXSOEBCFLV-UHFFFAOYSA-N | |
123066 | |
CC(C1=CC=CC=C1F)O |
C8H9FO | |
MFCD00014401 | |
1-2-fluorophenyl ethanol, 2-fluorophenylmethylcarbinol, 2-fluoro-alpha-methylbenzyl alcohol, 1-2-fluorophenyl ethan-1-ol, o-fluorophenylmethylcarbinol, 2-fluorophenyl methyl carbinol, 2-fluorophenylethanol, 1-m-fluorophenyl ethanol, 1-2'-fluorophenyl ethanol, fluorophenylmethylcarbinol | |
1-(2-fluorophenyl)ethanol |
Specifications
445-26-1 | |
MFCD00014401 | |
1-2-fluorophenyl ethanol, 2-fluorophenylmethylcarbinol, 2-fluoro-alpha-methylbenzyl alcohol, 1-2-fluorophenyl ethan-1-ol, o-fluorophenylmethylcarbinol, 2-fluorophenyl methyl carbinol, 2-fluorophenylethanol, 1-m-fluorophenyl ethanol, 1-2'-fluorophenyl ethanol, fluorophenylmethylcarbinol | |
SXFYVXSOEBCFLV-UHFFFAOYSA-N | |
1-(2-fluorophenyl)ethanol | |
123066 | |
95% |
C8H9FO | |
10 g | |
Immiscible in water. | |
CC(C1=CC=CC=C1F)O | |
140.157 | |
140.16 | |
1-(2-Fluorophenyl)ethanol |
Safety and Handling
EINECSNumber : 207-155-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only