missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Hexanediol 96.0+%, TCI America™
Supplier: TCI America H0688100ML
Specifications
1,2-Hexanediol | |
Colorless | |
C6H14O2 | |
100 mL | |
FHKSXSQHXQEMOK-LURJTMIESA-N | |
(2S)-hexane-1,2-diol | |
94335 | |
≥96.0% (GC) |
6920-22-5 | |
118°C | |
MFCD00010737 | |
1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol | |
CCCC[C@H](O)CO | |
118.18 | |
118.18 | |
Liquid |
Chemical Identifiers
6920-22-5 | |
118.18 | |
FHKSXSQHXQEMOK-LURJTMIESA-N | |
94335 | |
CCCC[C@H](O)CO |
C6H14O2 | |
MFCD00010737 | |
1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol | |
(2S)-hexane-1,2-diol |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : Yes