Learn More
1-(2-Hydroxyethyl)piperidine, 99%, Thermo Scientific Chemicals
$60.27 - $202.32
Chemical Identifiers
CAS | 3040-44-6 |
---|---|
Molecular Formula | C7H15NO |
Molecular Weight (g/mol) | 129.203 |
MDL Number | MFCD00006512 |
InChI Key | KZTWONRVIPPDKH-UHFFFAOYSA-N |
Synonym | 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol |
PubChem CID | 18232 |
ChEBI | CHEBI:61238 |
IUPAC Name | 2-piperidin-1-ylethanol |
SMILES | C1CCN(CC1)CCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2381622
|
Thermo Scientific Chemicals
B2381622 |
100 g |
Each for $60.27
|
|
|||||
AAB2381636
|
Thermo Scientific Chemicals
B2381636 |
500 g |
Each for $202.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3040-44-6 | |
129.203 | |
KZTWONRVIPPDKH-UHFFFAOYSA-N | |
18232 | |
2-piperidin-1-ylethanol |
C7H15NO | |
MFCD00006512 | |
2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol | |
CHEBI:61238 | |
C1CCN(CC1)CCO |
Specifications
3040-44-6 | |
0.977 | |
199°C to 202°C | |
Amine-like | |
1.4804 | |
100 g | |
103390 | |
2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol | |
C1CCN(CC1)CCO | |
129.203 | |
CHEBI:61238 | |
99% |
16°C | |
12.4 | |
83°C (181°F) | |
C7H15NO | |
MFCD00006512 | |
UN2735 | |
Air Sensitive | |
KZTWONRVIPPDKH-UHFFFAOYSA-N | |
2-piperidin-1-ylethanol | |
18232 | |
129.2 | |
1-(2-Hydroxyethyl)piperidine |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 221-244-6
RTECSNumber : TM8052000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only