Learn More
1-(2-Methoxyethyl)piperazine, 98%, Thermo Scientific Chemicals
$107.49 - $107.49
Chemical Identifiers
CAS | 13484-40-7 |
---|---|
Molecular Formula | C7H16N2O |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00191214 |
InChI Key | BMEMBBFDTYHTLH-UHFFFAOYSA-N |
Synonym | 1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine |
PubChem CID | 2734638 |
IUPAC Name | 1-(2-methoxyethyl)piperazine |
SMILES | COCCN1CCNCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC367110010
|
Thermo Scientific Chemicals
367110010 |
1 g | Glass bottle |
Each for $107.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13484-40-7 | |
144.21 | |
BMEMBBFDTYHTLH-UHFFFAOYSA-N | |
2734638 | |
COCCN1CCNCC1 |
C7H16N2O | |
MFCD00191214 | |
1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine | |
1-(2-methoxyethyl)piperazine |
Specifications
13484-40-7 | |
100.0 | |
0.9700g/mL | |
87°C | |
97.5% min. (GC) | |
C7H16N2O | |
MFCD00191214 | |
0.97 | |
BMEMBBFDTYHTLH-UHFFFAOYSA-N | |
1-(2-methoxyethyl)piperazine | |
2734638 | |
98% | |
1-(2-Methoxyethyl)piperazine, 98% |
97.5 | |
Colorless to Yellow | |
193°C to 194°C | |
Authentic | |
Glass bottle | |
1.4730 to 1.475 | |
1 g | |
1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine | |
COCCN1CCNCC1 | |
144.21 | |
144.21 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
RUO – Research Use Only