missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%, Thermo Scientific Chemicals

 

$184.96 - $426.59

Chemical Identifiers

CAS 521-24-4
Molecular Formula C10H5NaO5S
Molecular Weight (g/mol) 260.195
MDL Number MFCD00001700
InChI Key UBLXEEBHYISRFM-UHFFFAOYSA-M
Synonym 1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
PubChem CID 516996
IUPAC Name sodium;3,4-dioxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1050714
View Documents
Thermo Scientific Chemicals
A1050714
25 g
Each for $184.96
 
AAA1050722
View Documents
Thermo Scientific Chemicals
A1050722
100 g
Each for $426.59
 
Description

Description

Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.

Solubility
Soluble in water and acetone. Slightly soluble in alcohol and ethanol. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

521-24-4
260.195
UBLXEEBHYISRFM-UHFFFAOYSA-M
516996
C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]
C10H5NaO5S
MFCD00001700
1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
sodium;3,4-dioxonaphthalene-1-sulfonate

Specifications

521-24-4
C10H5NaO5S
25 g
14,8646
Soluble in water and acetone; Slightly soluble in alcohol and ethanol; Insoluble in chloroform.
C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]
260.195
260.2
1,2-Naphthoquinone-4-sulfonic acid sodium salt
∼289°C (decomposition)
MFCD00001700
4169788
1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
UBLXEEBHYISRFM-UHFFFAOYSA-M
sodium;3,4-dioxonaphthalene-1-sulfonate
516996
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-308-9

RTECSNumber : QK1490000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only