Learn More
1,2-Pentanediol, 96%
CAS: 5343-92-0 | C5H12O2 | 104.15 g/mol
$204.94 - $204.94
Chemical Identifiers
| CAS | 5343-92-0 |
|---|---|
| Molecular Formula | C5H12O2 |
| Molecular Weight (g/mol) | 104.15 |
| MDL Number | MFCD00010736 |
| InChI Key | WCVRQHFDJLLWFE-UHFFFAOYNA-N |
| Synonym | 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa |
| PubChem CID | 93000 |
| IUPAC Name | pentane-1,2-diol |
| SMILES | CCCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC123290500
|
Thermo Scientific Chemicals
123290500 |
50 mL | Glass bottle |
Each for $204.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5343-92-0 | |
| 104.15 | |
| WCVRQHFDJLLWFE-UHFFFAOYNA-N | |
| 93000 | |
| CCCC(O)CO |
| C5H12O2 | |
| MFCD00010736 | |
| 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa | |
| pentane-1,2-diol |
Specifications
| 5343-92-0 | |
| 100.0 | |
| 0.9710g/mL | |
| 105°C | |
| 95% min. (GC) | |
| C5H12O2 | |
| CH3(CH2)2CH(OH)CH2OH | |
| 50 mL | |
| 0.971 | |
| Solubility in water: miscible. | |
| CCCC(O)CO | |
| 104.15 | |
| 104.15 | |
| Liquid |
| 96.0 | |
| Colorless to Yellow | |
| 206.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4387 to 1.4407 | |
| MFCD00010736 | |
| 01,II,548 | |
| 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa | |
| WCVRQHFDJLLWFE-UHFFFAOYNA-N | |
| pentane-1,2-diol | |
| 93000 | |
| 96% | |
| 1, 2-Pentanediol, 97% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 226-285-3
RTECSNumber : SA0455000
TSCA : TSCA
RUO – Research Use Only