Learn More
1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 26388-13-6 |
|---|---|
| Molecular Formula | C14H12O2 |
| Molecular Weight (g/mol) | 212.25 |
| MDL Number | MFCD08271961 |
| InChI Key | KPBCVVSDGJBODL-UHFFFAOYSA-N |
| Synonym | 1-2-phenoxyphenyl ethanone, 2-acetyldiphenyl ether, 1-acetyl-2-phenoxybenzene, ethanone, 1-2-phenoxyphenyl |
| PubChem CID | 10703750 |
| IUPAC Name | 1-(2-phenoxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC=CC=C1OC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
CC49928CB
|
Thermo Scientific Chemicals
CC49928CB |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC49928DA
|
Thermo Scientific Chemicals
CC49928DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC49928DE
|
Thermo Scientific Chemicals
CC49928DE |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 26388-13-6 | |
| 212.25 | |
| KPBCVVSDGJBODL-UHFFFAOYSA-N | |
| 10703750 | |
| CC(=O)C1=CC=CC=C1OC1=CC=CC=C1 |
| C14H12O2 | |
| MFCD08271961 | |
| 1-2-phenoxyphenyl ethanone, 2-acetyldiphenyl ether, 1-acetyl-2-phenoxybenzene, ethanone, 1-2-phenoxyphenyl | |
| 1-(2-phenoxyphenyl)ethanone |
Specifications
| 26388-13-6 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD08271961 | |
| 1-2-phenoxyphenyl ethanone, 2-acetyldiphenyl ether, 1-acetyl-2-phenoxybenzene, ethanone, 1-2-phenoxyphenyl | |
| CC(=O)C1=CC=CC=C1OC1=CC=CC=C1 | |
| 212.25 | |
| 212.25 | |
| 1-(2-Phenoxyphenyl)ethanone |
| 96.5 | |
| 97% | |
| C14H12O2 | |
| 250mg | |
| KPBCVVSDGJBODL-UHFFFAOYSA-N | |
| 1-(2-phenoxyphenyl)ethanone | |
| 10703750 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning