Learn More
1,2-Phenylenediacetic acid, 98+%
CAS: 7500-53-0 | C10H10O4 | 194.186 g/mol
$38.12 - $128.15
Chemical Identifiers
| CAS | 7500-53-0 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.186 |
| MDL Number | MFCD00004329 |
| InChI Key | MMEDJBFVJUFIDD-UHFFFAOYSA-N |
| Synonym | 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid |
| PubChem CID | 24123 |
| IUPAC Name | 2-[2-(carboxymethyl)phenyl]acetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1385106
|
Thermo Scientific Chemicals
A1385106 |
5 g |
Each for $38.12
|
|
|||||
|
AAA1385114
|
Thermo Scientific Chemicals
A1385114 |
25 g |
Each for $128.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7500-53-0 | |
| 194.186 | |
| MMEDJBFVJUFIDD-UHFFFAOYSA-N | |
| 24123 | |
| C1=CC=C(C(=C1)CC(=O)O)CC(=O)O |
| C10H10O4 | |
| MFCD00004329 | |
| 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid | |
| 2-[2-(carboxymethyl)phenyl]acetic acid |
Specifications
| 7500-53-0 | |
| Odorless | |
| MFCD00004329 | |
| 2616747 | |
| MMEDJBFVJUFIDD-UHFFFAOYSA-N | |
| 2-[2-(carboxymethyl)phenyl]acetic acid | |
| 24123 | |
| ≥98% |
| 150°C to 152°C | |
| C10H10O4 | |
| 5 g | |
| 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid | |
| C1=CC=C(C(=C1)CC(=O)O)CC(=O)O | |
| 194.186 | |
| 194.19 | |
| 1,2-Phenylenediacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-355-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only