Learn More
1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 165900250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(2-Pyridyl)piperazine | |
97.5 | |
Colorless to Yellow | |
120°C to 122°C (2.0 mmHg) | |
97.5% min. (GC) | |
C9H13N3 | |
25 g | |
1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine | |
GZRKXKUVVPSREJ-UHFFFAOYSA-N | |
1-pyridin-2-ylpiperazine | |
94459 | |
98% |
34803-66-2 | |
100.0 | |
1.0700g/mL | |
Authentic | |
Glass bottle | |
1.5930 to 1.595 | |
1.07 | |
Solubility in water: >500g/L (20 c). Other solubilities: >50g/100mL methanol- >50g/100mL ethanol (20 c),>50g/100mL 2-propanol- >50g/100mL aceton (20 c),>50g/100mL n,n-dimethylformamide (20 c),>50g/100mL methylen chloride | |
C1CN(CCN1)C2=CC=CC=N2 | |
163.22 | |
163.22 | |
Liquid |
Chemical Identifiers
34803-66-2 | |
163.22 | |
1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine | |
1-pyridin-2-ylpiperazine |
C9H13N3 | |
GZRKXKUVVPSREJ-UHFFFAOYSA-N | |
94459 | |
C1CN(CCN1)C2=CC=CC=N2 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 252-220-3
RUO – Research Use Only