missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-2-methyleneindoline, 95%

CAS: 118-12-7 | C12H15N | 173.259 g/mol

$123.22 - $467.81

Chemical Identifiers

CAS 118-12-7
Molecular Formula 173.259
Molecular Weight (g/mol) 173.259
MDL Number MFCD00005813
InChI Key ZTUKGBOUHWYFGC-UHFFFAOYSA-N
Synonym 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
PubChem CID 8351
IUPAC Name 1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=CC=CC=C21)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1674118
View Documents
Thermo Scientific Chemicals
A1674118
50 g N/A
Only null left
 
AAA1674130
View Documents
Thermo Scientific Chemicals
A1674130
250 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

118-12-7
173.259
ZTUKGBOUHWYFGC-UHFFFAOYSA-N
8351
CC1(C(=C)N(C2=CC=CC=C21)C)C
C12H15N
MFCD00005813
1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
1,3,3-trimethyl-2-methylideneindole

Specifications

118-12-7
0.979
101°C (213°F)
1.578
50 g
1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
CC1(C(=C)N(C2=CC=CC=C21)C)C
173.259
173.26
1,3,3-Trimethyl-2-methyleneindoline
-10°C
247°C to 248°C
C12H15N
MFCD00005813
131162
ZTUKGBOUHWYFGC-UHFFFAOYSA-N
1,3,3-trimethyl-2-methylideneindole
8351
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332

EINECSNumber : 204-235-1

RTECSNumber : NM1926500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only