Learn More
1,3,5,7-Cyclooctatetraene, 98%, stabilized
CAS: 629-20-9 | C8H8 | 104.15 g/mol
$268.13 - $591.12
Chemical Identifiers
| CAS | 629-20-9 |
|---|---|
| Molecular Formula | C8H8 |
| Molecular Weight (g/mol) | 104.15 |
| MDL Number | MFCD00004161 |
| InChI Key | KDUIUFJBNGTBMD-DLMDZQPMSA-N |
| Synonym | 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n |
| PubChem CID | 637866 |
| IUPAC Name | cyclooctatetraene |
| SMILES | C1=C/C=C\C=C/C=C\1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC160570025
|
Thermo Scientific Chemicals
160570025 |
2.5 g | Glass bottle |
N/A
|
|
||||
|
AC160570100
|
Thermo Scientific Chemicals
160570100 |
10 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 629-20-9 | |
| 104.15 | |
| KDUIUFJBNGTBMD-DLMDZQPMSA-N | |
| 637866 | |
| C1=C/C=C\C=C/C=C\1 |
| C8H8 | |
| MFCD00004161 | |
| 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n | |
| cyclooctatetraene |
Specifications
| 629-20-9 | |
| 100.0 | |
| Brown-Yellow to Yellow | |
| 142°C to 143°C | |
| Authentic | |
| Glass bottle | |
| 1.5365 to 1.5385 | |
| 2.5 g | |
| 0.92 | |
| 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n | |
| KDUIUFJBNGTBMD-DLMDZQPMSA-N | |
| cyclooctatetraene | |
| 0.1% hydroquinone | |
| 104.15 | |
| Liquid |
| 97.5 | |
| -27°C | |
| 0.9200g/mL | |
| 22°C | |
| 97.5% min. (GC) | |
| C8H8 | |
| MFCD00004161 | |
| 05, I, 228 | |
| 15, 2731 | |
| Solubility in water: immiscible | |
| C1=C/C=C\C=C/C=C\1 | |
| 104.15 | |
| 637866 | |
| 98% | |
| 1,3,5,7-Cyclooctatetraene, Stabilized |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti
GHS Signal Word: Danger
EINECSNumber : 211-080-3
RUO – Research Use Only