Learn More
1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
$42.04 - $332.44
Chemical Identifiers
CAS | 4422-95-1 |
---|---|
Molecular Formula | C9H3Cl3O3 |
Molecular Weight (g/mol) | 265.47 |
MDL Number | MFCD00000679 |
InChI Key | UWCPYKQBIPYOLX-UHFFFAOYSA-N |
Synonym | trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride |
PubChem CID | 78138 |
SMILES | ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0725506
|
Thermo Scientific Chemicals
L0725506 |
5 g |
Each for $42.04
|
|
|||||
AAL0725514
|
Thermo Scientific Chemicals
L0725514 |
25 g |
Each for $114.58
|
|
|||||
AAL0725522
|
Thermo Scientific Chemicals
L0725522 |
100 g |
Each for $332.44
|
|
|||||
Description
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.
Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.
Chemical Identifiers
4422-95-1 | |
265.47 | |
UWCPYKQBIPYOLX-UHFFFAOYSA-N | |
78138 |
C9H3Cl3O3 | |
MFCD00000679 | |
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride | |
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O |
Specifications
4422-95-1 | |
180°C (16 mmHg) | |
C9H3Cl3O3 | |
5 g | |
2940936 | |
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride | |
UWCPYKQBIPYOLX-UHFFFAOYSA-N | |
benzene-1,3,5-tricarbonyl trichloride | |
78138 | |
≥98% |
34°C to 38°C | |
>110°C (230°F) | |
MFCD00000679 | |
UN3261 | |
Moisture sensitive | |
Soluble in chloroform(soluble10%,clear to very slightly hazy,colorless to faintly yellow) and toluene. Solubility in water: Reacts. | |
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O | |
265.47 | |
265.48 | |
1,3,5-Benzenetricarbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 224-594-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only