missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Benzenetricarboxylic acid chloride, 98%, Thermo Scientific Chemicals

$59.67 - $354.51

Chemical Identifiers

CAS 4422-95-1
Molecular Formula C9H3Cl3O3
Molecular Weight (g/mol) 265.47
MDL Number MFCD00000679
InChI Key UWCPYKQBIPYOLX-UHFFFAOYSA-N
Synonym trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
PubChem CID 78138
IUPAC Name benzene-1,3,5-tricarbonyl chloride
SMILES ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC164370050
SDS View Documents
Thermo Scientific Chemicals
164370050
5 g Glass bottle
Each for $59.67
 
AC164371000
SDS View Documents
Thermo Scientific Chemicals
164371000
100 g Glass bottle
Each for $354.51
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4422-95-1
265.47
UWCPYKQBIPYOLX-UHFFFAOYSA-N
78138
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
C9H3Cl3O3
MFCD00000679
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
benzene-1,3,5-tricarbonyl chloride

Specifications

4422-95-1
100.0
Colorless to Yellow
>112°C
97.5 to 102.5% (ex Cl) (Argentometry)
C9H3Cl3O3
C6H3(COCl)3
5 g
trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
UWCPYKQBIPYOLX-UHFFFAOYSA-N
benzene-1,3,5-tricarbonyl chloride
78138
98%
1, 3, 5-Benzenetricarboxylic acid chloride
97.5
34.5°C to 36°C
180°C (16.0 mmHg)
Authentic
Glass bottle
1.5945 to 1.5965
MFCD00000679
09, IV, 3748
Solubility in water: decomposes
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
265.47
265.47
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-594-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only