missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol, 98%, Thermo Scientific Chemicals
$53.33 - $595.33
Chemical Identifiers
CAS | 368-63-8 |
---|---|
Molecular Formula | C10H8F6O |
Molecular Weight (g/mol) | 258.16 |
MDL Number | MFCD01632521,MFCD03093011 |
InChI Key | MMSCIQKQJVBPIR-UHFFFAOYNA-N |
Synonym | 1-3,5-bis trifluoromethyl phenyl ethanol, 3,5-bis trifluoromethyl phenyl ethanol, 1-3,5-bis trifluoromethyl phenyl ethan-1-ol, alfa-methyl-3,5-ditrifluoromethylbenzyl alcohol, pubchem4672, pubchem14254, 1-3,5-bis-trifluoromethylphenyl ethanol, acmc-209vt9, benzenemethanol, alpha-methyl-3,5-bis trifluoromethyl |
PubChem CID | 6425739 |
IUPAC Name | 1-[3,5-bis(trifluoromethyl)phenyl]ethanol |
SMILES | CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3197203
|
Thermo Scientific Chemicals
H3197203 |
1 g |
Each for $53.33
|
|
|||||
AAH3197206
|
Thermo Scientific Chemicals
H3197206 |
5 g |
Each for $178.05
|
|
|||||
AAH3197214
|
Thermo Scientific Chemicals
H3197214 |
25 g |
Each for $595.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
368-63-8 | |
258.16 | |
MMSCIQKQJVBPIR-UHFFFAOYNA-N | |
6425739 | |
CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
C10H8F6O | |
MFCD01632521,MFCD03093011 | |
1-3,5-bis trifluoromethyl phenyl ethanol, 3,5-bis trifluoromethyl phenyl ethanol, 1-3,5-bis trifluoromethyl phenyl ethan-1-ol, alfa-methyl-3,5-ditrifluoromethylbenzyl alcohol, pubchem4672, pubchem14254, 1-3,5-bis-trifluoromethylphenyl ethanol, acmc-209vt9, benzenemethanol, alpha-methyl-3,5-bis trifluoromethyl | |
1-[3,5-bis(trifluoromethyl)phenyl]ethanol |
Specifications
368-63-8 | |
C10H8F6O | |
1 g | |
MMSCIQKQJVBPIR-UHFFFAOYNA-N | |
1-[3,5-bis(trifluoromethyl)phenyl]ethanol | |
6425739 | |
98% |
70°C to 74°C | |
MFCD01632521,MFCD03093011 | |
1-3,5-bis trifluoromethyl phenyl ethanol, 3,5-bis trifluoromethyl phenyl ethanol, 1-3,5-bis trifluoromethyl phenyl ethan-1-ol, alfa-methyl-3,5-ditrifluoromethylbenzyl alcohol, pubchem4672, pubchem14254, 1-3,5-bis-trifluoromethylphenyl ethanol, acmc-209vt9, benzenemethanol, alpha-methyl-3,5-bis trifluoromethyl | |
CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F | |
258.16 | |
258.16 | |
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only