Learn More
1-(3,5-Difluoro-2-hydroxyphenyl)ethan-1-one, Thermo Scientific™
$49.26 - $394.06
Chemical Identifiers
CAS | 140675-42-9 |
---|---|
Molecular Formula | C8H6F2O2 |
Molecular Weight (g/mol) | 172.131 |
MDL Number | MFCD00042476 |
InChI Key | MCDJUVXLLXTCFP-UHFFFAOYSA-N |
Synonym | 1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone |
PubChem CID | 2736976 |
IUPAC Name | 1-(3,5-difluoro-2-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=CC(=C1O)F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
BTB04131DA
|
Thermo Scientific Chemicals
BTB04131DA |
1 g | Amber Glass Bottle |
N/A
|
|
|||||
BTB04131EA
|
Thermo Scientific Chemicals
BTB04131EA |
10 g | Amber Glass Bottle |
N/A
|
|
|||||
Chemical Identifiers
140675-42-9 | |
172.131 | |
MCDJUVXLLXTCFP-UHFFFAOYSA-N | |
2736976 | |
CC(=O)C1=CC(=CC(=C1O)F)F |
C8H6F2O2 | |
MFCD00042476 | |
1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone | |
1-(3,5-difluoro-2-hydroxyphenyl)ethanone |
Specifications
140675-42-9 | |
100.0 | |
Amber Glass Bottle | |
MFCD00042476 | |
1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone | |
CC(=O)C1=CC(=CC(=C1O)F)F | |
172.131 | |
172.13 | |
1-(3, 5-Difluoro-2-hydroxyphenyl)ethan-1-one |
96.5 | |
97% | |
C8H6F2O2 | |
1 g | |
MCDJUVXLLXTCFP-UHFFFAOYSA-N | |
1-(3,5-difluoro-2-hydroxyphenyl)ethanone | |
2736976 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.