Learn More
1,3,5-Tri(4-carboxyphenyl)benzene, 97%
CAS: 50446-44-1 | C27H18O6 | 438.44 g/mol
$220.66 - $760.85
Chemical Identifiers
| CAS | 50446-44-1 |
|---|---|
| Molecular Formula | C27H18O6 |
| Molecular Weight (g/mol) | 438.44 |
| MDL Number | MFCD10000888 |
| InChI Key | SATWKVZGMWCXOJ-UHFFFAOYSA-N |
| Synonym | 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid |
| PubChem CID | 10694305 |
| IUPAC Name | 4-[3,5-bis(4-carboxyphenyl)phenyl]benzoic acid |
| SMILES | OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6008703
|
Thermo Scientific Chemicals
H6008703 |
1 g |
Each for $220.66
|
|
|||||
|
AAH6008706
|
Thermo Scientific Chemicals
H6008706 |
5 g |
Each for $760.85
|
|
|||||
Description
1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.
Solubility
Soluble in tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents, metals, amines, strong acids, acid chlorides, alcohols, peroxides, permanganates, strong reducing agents, soluble carbonates, phosphates, phosphorus halides and hydroxides.
Chemical Identifiers
| 50446-44-1 | |
| 438.44 | |
| SATWKVZGMWCXOJ-UHFFFAOYSA-N | |
| 10694305 | |
| OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O |
| C27H18O6 | |
| MFCD10000888 | |
| 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid | |
| 4-[3,5-bis(4-carboxyphenyl)phenyl]benzoic acid |
Specifications
| 50446-44-1 | |
| C27H18O6 | |
| 1 g | |
| 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid | |
| SATWKVZGMWCXOJ-UHFFFAOYSA-N | |
| 4-[3,5-bis(4-carboxyphenyl)phenyl]benzoic acid | |
| 10694305 | |
| 97% |
| 322°C to 327°C | |
| MFCD10000888 | |
| UN3077 | |
| Soluble in tetrahydrofuran. | |
| OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O | |
| 438.44 | |
| 438.43 | |
| 1,3,5-Tri(4-carboxyphenyl)benzene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only