Learn More
1,3,5-Tri-n-propylhexahydro-1,3,5-triazine, 96%, Thermo Scientific™
$49.00 - $136.44
Chemical Identifiers
CAS | 13036-81-2 |
---|---|
Molecular Formula | C12H27N3 |
Molecular Weight (g/mol) | 213.37 |
MDL Number | MFCD00040461 |
InChI Key | DPHBVWJGMBYPMK-UHFFFAOYSA-N |
Synonym | 1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine |
PubChem CID | 83068 |
IUPAC Name | 1,3,5-tripropyl-1,3,5-triazinane |
SMILES | CCCN1CN(CCC)CN(CCC)C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH64078MD
|
Thermo Scientific Chemicals
H64078MD |
250 mg |
Each for $49.00
|
|
|||||
AAH6407803
|
Thermo Scientific Chemicals
H6407803 |
1 g |
Each for $136.44
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13036-81-2 | |
213.37 | |
DPHBVWJGMBYPMK-UHFFFAOYSA-N | |
83068 | |
CCCN1CN(CCC)CN(CCC)C1 |
C12H27N3 | |
MFCD00040461 | |
1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine | |
1,3,5-tripropyl-1,3,5-triazinane |
Specifications
13036-81-2 | |
0.880 g/mL | |
C12H27N3 | |
MFCD00040461 | |
111423 | |
DPHBVWJGMBYPMK-UHFFFAOYSA-N | |
1,3,5-tripropyl-1,3,5-triazinane | |
83068 | |
96% | |
1,3,5-Tri-n-propylhexahydro-1,3,5-triazine |
Colorless | |
94°C (0.7 mmHg) | |
1.466 | |
250 mg | |
1,3,5-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-s-triazine, s-triazine, hexahydro-1,3,5-tripropyl, hexahydro-1,3,5-tripropyl-1,3,5-triazine, #, 1,3,5-tri-n-propyl hexahydro-s-triazine, 1,3,5-tripropylhexahydro-1,3,5-triazine, 1,3,5-tri-n-propylhexahydro-1,3,5-triazine | |
CCCN1CN(CCC)CN(CCC)C1 | |
213.37 | |
213.37 | |
Liquid |
Safety and Handling
GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 235-904-6
TSCA : Yes
Recommended Storage : Ambient temperatures