missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Triaza-7-phosphaadamantane, 97+%, Thermo Scientific Chemicals

$150.85 - $693.22

Chemical Identifiers

CAS 53597-69-6
Molecular Formula C6H12N3P
Molecular Weight (g/mol) 157.157
MDL Number MFCD00154905
InChI Key FXXRPTKTLVHPAR-UHFFFAOYSA-N
Synonym 1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
PubChem CID 143061
SMILES C1N2CN3CN1CP(C2)C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6678503
View Documents
Thermo Scientific Chemicals
H6678503
1 g
Each for $150.85
 
AAH6678506
View Documents
Thermo Scientific Chemicals
H6678506
5 g
Each for $693.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53597-69-6
157.157
FXXRPTKTLVHPAR-UHFFFAOYSA-N
143061
C6H12N3P
MFCD00154905
1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
C1N2CN3CN1CP(C2)C3

Specifications

53597-69-6
White
MFCD00154905
Hygroscopic
FXXRPTKTLVHPAR-UHFFFAOYSA-N
157.157
157.15
Solid
244°C to 250°C
C6H12N3P
1 g
1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
C1N2CN3CN1CP(C2)C3
143061
≥97%
1,3,5-Triaza-7-phosphaadamantane
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only