missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Triazine 98.0+%, TCI America™
$138.25 - $1167.22
Chemical Identifiers
CAS | 290-87-9 |
---|---|
Molecular Formula | C3H3N3 |
Molecular Weight (g/mol) | 81.08 |
MDL Number | MFCD00006044 |
InChI Key | JIHQDMXYYFUGFV-UHFFFAOYSA-N |
Synonym | s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t |
PubChem CID | 9262 |
ChEBI | CHEBI:30259 |
IUPAC Name | 1,3,5-triazine |
SMILES | C1=NC=NC=N1 |
Chemical Identifiers
290-87-9 | |
81.08 | |
JIHQDMXYYFUGFV-UHFFFAOYSA-N | |
9262 | |
1,3,5-triazine |
C3H3N3 | |
MFCD00006044 | |
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t | |
CHEBI:30259 | |
C1=NC=NC=N1 |
Specifications
290-87-9 | |
White-Yellow | |
C3H3N3 | |
1 g | |
JIHQDMXYYFUGFV-UHFFFAOYSA-N | |
1,3,5-triazine | |
9262 | |
81.08 | |
Crystalline Powder |
81°C | |
114°C | |
MFCD00006044 | |
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t | |
C1=NC=NC=N1 | |
81.08 | |
CHEBI:30259 | |
≥98.0% (GC) | |
1,3,5-Triazine |
Safety and Handling
RTECSNumber : XY2957000
TSCA : Yes