missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Tribenzylhexahydro-1,3,5-triazine, 98+%, Thermo Scientific Chemicals

$36.36 - $121.27

Chemical Identifiers

CAS 2547-66-2
Molecular Formula C24H27N3
Molecular Weight (g/mol) 357.50
MDL Number MFCD00014599
InChI Key VWVZIRPJPFJGFE-UHFFFAOYSA-N
Synonym 1,3,5-tribenzylhexahydro-1,3,5-triazine, 1,3,5-tribenzylhexahydro-s-triazine, 1,3,5-triazine, hexahydro-1,3,5-tris phenylmethyl, 1,3,5-trisbenzyl-1,3,5-triazaperhydroine, acmc-1cmek, 1,3,5-tribenzyl-1,3,5 triazinane, 1,3,5-triazine,hexahydro-1,3,5-tris phenylmethyl
PubChem CID 75685
IUPAC Name 1,3,5-tribenzyl-1,3,5-triazinane
SMILES C(N1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2596406
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H2596406
5 g N/A
 
AAH2596414
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H2596414
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2547-66-2
357.50
VWVZIRPJPFJGFE-UHFFFAOYSA-N
75685
C(N1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1)C1=CC=CC=C1
C24H27N3
MFCD00014599
1,3,5-tribenzylhexahydro-1,3,5-triazine, 1,3,5-tribenzylhexahydro-s-triazine, 1,3,5-triazine, hexahydro-1,3,5-tris phenylmethyl, 1,3,5-trisbenzyl-1,3,5-triazaperhydroine, acmc-1cmek, 1,3,5-tribenzyl-1,3,5 triazinane, 1,3,5-triazine,hexahydro-1,3,5-tris phenylmethyl
1,3,5-tribenzyl-1,3,5-triazinane

Specifications

2547-66-2
100°C (0.005 mmHg)
C24H27N3
5 g
VWVZIRPJPFJGFE-UHFFFAOYSA-N
1,3,5-tribenzyl-1,3,5-triazinane
75685
≥98%
50°C to 51°C
113°C (235°F)
MFCD00014599
1,3,5-tribenzylhexahydro-1,3,5-triazine, 1,3,5-tribenzylhexahydro-s-triazine, 1,3,5-triazine, hexahydro-1,3,5-tris phenylmethyl, 1,3,5-trisbenzyl-1,3,5-triazaperhydroine, acmc-1cmek, 1,3,5-tribenzyl-1,3,5 triazinane, 1,3,5-triazine,hexahydro-1,3,5-tris phenylmethyl
C(N1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1)C1=CC=CC=C1
357.50
357.49
1,3,5-Tribenzylhexahydro-1,3,5-triazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-831-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only